Learn More
Tocris Bioscience™ Licarbazepine
Active metabolite of oxcarbazepine (Cat. No. 3864)
POA
Specifications
Purity >99%
View More Specs
Description
Description
Active metabolite of oxcarbazepine (Cat. No. 3864). Produces dose-dependent inhibition of glutamatergic excitatory postsynaptic potentials (EPSPs). Displays anticonvulsant activity; exhibits minor potentiation of GABAA receptor currents.
Specifications
Specifications
>99%