missing translation for 'onlineSavingsMsg'
Learn More

Liarozole hydrochloride, Tocris Bioscience™

Blocks retinoic acid metabolism

Brand:  Tocris (Bio-Techne) 270550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431764

  • £764.40 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytochrome P450 inhibitor. Inhibits several cytochrome P450 enzymes including aromatase (CYP19) and retinoic acid 4-hydroxylase (CYP26). Blocks retinoic acid metabolism (retinoic acid metabolism blocking agent, RAMBA). Displays antitumor activity against androgen-dependent and independent rat prostate carcinomas.
Specifications

Chemical Identifiers

145858-50-0
345.227
liarozole hydrochloride, liarozole hcl, liarozole hydrochloride usan, dsstox_cid_26817, dsstox_rid_81929, dsstox_gsid_46817, 1h-benzimidazole,6-3-chlorophenyl-1h-imidazol-1-ylmethyl, 5-3-chlorophenyl imidazol-1-yl methyl-1h-1,3-benzodiazole hydrochloride, acmc-20b0ri
6-[(3-chlorophenyl)-imidazol-1-ylmethyl]-1H-benzimidazole;hydrochloride
C17H14Cl2N4
OTSZCHORPMQCBZ-UHFFFAOYSA-N
71454
C1=CC(=CC(=C1)Cl)C(C2=CC3=C(C=C2)N=CN3)N4C=CN=C4.Cl

Specifications

50mg
145858-50-0
liarozole hydrochloride, liarozole hcl, liarozole hydrochloride usan, dsstox_cid_26817, dsstox_rid_81929, dsstox_gsid_46817, 1h-benzimidazole,6-3-chlorophenyl-1h-imidazol-1-ylmethyl, 5-3-chlorophenyl imidazol-1-yl methyl-1h-1,3-benzodiazole hydrochloride, acmc-20b0ri
C1=CC(=CC(=C1)Cl)C(C2=CC3=C(C=C2)N=CN3)N4C=CN=C4.Cl
345.227
345.23
Liarozole hydrochloride
C17H14Cl2N4
OTSZCHORPMQCBZ-UHFFFAOYSA-N
6-[(3-chlorophenyl)-imidazol-1-ylmethyl]-1H-benzimidazole;hydrochloride
71454
>99%
Videos
Documents