missing translation for 'onlineSavingsMsg'
Learn More

LG 100754, Tocris Bioscience™

RXR:PPAR agonist

Brand:  Tocris (Bio-Techne) 383110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443674

  • £163.17 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Novel RXR:PPARγ agonist; sensitizes PPARγ by enhancing its ligand binding activity. Also activates RXR:RAR and RXR:PPARα heterodimers in cotransfection assays. Displays selectivity over other permissive heterodimers such as RXR:LXRα and RXR:BAR/FXR. Exhibits antidiabetic properties in vivo.
Specifications

Chemical Identifiers

180713-37-5
396.571
(2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
(2E,4E,6E)-3-methyl-7-(5,5,8,8-tetramethyl-3-propoxy-6,7-dihydronaphthalen-2-yl)octa-2,4,6-trienoic acid
C26H36O3
HNODNXQAYXJFMQ-ZHUZXUJZSA-N
12020728
CCCOC1=CC2=C(C=C1C(=CC=CC(=CC(=O)O)C)C)C(CCC2(C)C)(C)C

Specifications

10mg
180713-37-5
(2E,4E,6Z)-3-Methyl-7-(5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-propoxy-3-naphthalenyl)-2,4,6-octatrienoic acid
CCCOC1=CC2=C(C=C1C(=CC=CC(=CC(=O)O)C)C)C(CCC2(C)C)(C)C
396.571
396.56
LG 100754
C26H36O3
HNODNXQAYXJFMQ-ZHUZXUJZSA-N
(2E,4E,6E)-3-methyl-7-(5,5,8,8-tetramethyl-3-propoxy-6,7-dihydronaphthalen-2-yl)octa-2,4,6-trienoic acid
12020728
>99%
Videos
Documents