missing translation for 'onlineSavingsMsg'
Learn More

Lewis Y Mouse, Clone: A70-A/A9, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio MSM2-1412-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941388

  • £379.00 / 100µg

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Lewis Y
Cell membrane, cytoplasmic and extracellular
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
Live Ls174T cells (human colon carcinoma cell line)
Lung, Colon, Bladder or Hepatocellular Carcinoma
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Immunohistochemistry
A70-A/A9
Unconjugated
Mouse
IgG1 kappa
Protein A/G Purified
RUO
Primary
Videos
SDS
Documents