missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levulinic Acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 33.0°C to 35.0°C |
Color | Yellow |
Density | 1.1300g/mL |
Boiling Point | 245.0°C to 246.0°C |
Flash Point | 98°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Plastic drum |
Linear Formula | CH3COCH2CH2CO2H |
Refractive Index | 1.4396 to 1.4426 |
Show More |
Safety and Handling
Product Identifier
- Levulinic acid
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'