missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam, Tocris Bioscience™

Antiepileptic; binds SV2A

Brand:  Tocris (Bio-Techne) 283910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13431924

  • £110.25 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Antiepileptic that displays distinctive properties from conventional antiepileptic drugs. Displays potent seizure protection in animal models of chronic epilepsy but lacks activity in acute seizure models. Binds synaptic vesicle protein 2A (SV2A) and inhibits Na+-dependent Cl-/HCO3- exchange.

Deuterated analog also available.

Specifications

Chemical Identifiers

102767-28-2
170.212
levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
CHEBI:6437
CCC(C(=O)N)N1CCCC1=O
C8H14N2O2
HPHUVLMMVZITSG-LURJTMIESA-N
5284583
(2S)-2-(2-oxopyrrolidin-1-yl)butanamide

Specifications

10mg
102767-28-2
levetiracetam, keppra, s-2-2-oxopyrrolidin-1-yl butanamide, keppra xr, 2s-2-2-oxopyrrolidin-1-yl butanamide, levetiracetamum, ucb-l 059, ucb l059, levetiractam, levroxa
CCC(C(=O)N)N1CCCC1=O
170.212
CHEBI:6437
>99%
Levetiracetam
C8H14N2O2
HPHUVLMMVZITSG-LURJTMIESA-N
(2S)-2-(2-oxopyrrolidin-1-yl)butanamide
5284583
170.21
Videos
Documents