missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leupeptin, 90%, synthetic
56.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
49.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
49.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Product Type | Leupeptin |
Formula Weight | 475.6 |
Solubility | Solubility in water: soluble in water |
Form | Powder |
Quantity | 5 mg |
Percent Purity | 90% |
Chemical Name or Material | Leupeptin |
Safety and Handling
Product Identifier
- Leupeptin
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'