missing translation for 'onlineSavingsMsg'
Learn More

Lercanidipine hydrochloride, Tocris Bioscience™

CaV1.x blocker

Brand:  Tocris (Bio-Techne) 295910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452134

  • £99.96 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

L-type Ca2+ channel blocker that displays higher vascular selectivity than felodipine (Cat. No. 2960). Causes peripheral vasodilation with only weak negative inotropic activity. Antihypertensive.
Specifications

Chemical Identifiers

132866-11-6
648.197
lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen
5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
C36H42ClN3O6
WMFYOYKPJLRMJI-UHFFFAOYSA-N
157917
CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl

Specifications

10mg
132866-11-6
lercanidipine hydrochloride, lercanidipine hcl, cardiovasc, carmen, corifeo, lerzam, lercadip, lercapin, lercaton, lerkamen
CC1=C(C(C(=C(N1)C)C(=O)OC(C)(C)CN(C)CCC(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl
648.197
648.19
Lercanidipine hydrochloride
C36H42ClN3O6
WMFYOYKPJLRMJI-UHFFFAOYSA-N
5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
157917
>99%
Videos
Documents