missing translation for 'onlineSavingsMsg'
Learn More

4-Methylquinoline, 99%, Thermo Scientific Chemicals

Catalog Number 10246030
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
491-35-0
C10H9N
143.19
MFCD00006784
MUDSDYNRBDKLGK-UHFFFAOYSA-N
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
10285
CHEBI:48983
4-methylquinoline
CC1=CC=NC2=CC=CC=C12
This item is not returnable. View return policy
491-35-0
C10H9N
143.19
MFCD00006784
MUDSDYNRBDKLGK-UHFFFAOYSA-N
lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
10285
CHEBI:48983
4-methylquinoline
CC1=CC=NC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 9.0°C to 10.0°C
Density 1.0800g/mL
Boiling Point 261.0°C to 263.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6180 to 1.6200
Quantity 5 g
Beilstein 20, 395
Merck Index 15, 5496
Specific Gravity 1.08
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene and ether
Formula Weight 143.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylquinoline, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methylquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'