Learn More
LENG8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559847
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PERELKKQKR AARFQHGHSR RLRLEPLVLQ MSSLESSGAD PDWQELQIVG TCPDITKHYL RLTCAPDPST VRPVAVLKKS LCMVKCHWKE KQDYAFACEQ MKSIRQDLTV Q Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
LENG8 (Leukocyte Receptor Cluster Member 8) is a Protein Coding gene.Specifications
LENG8 | |
Polyclonal | |
Unconjugated | |
LENG8 | |
AW049671; KIAA1932; LENG8; leukocyte receptor cluster (LRC) member 8; leukocyte receptor cluster member 8; Leukocyte receptor cluster member 8 homolog; MGC40108; pp13842 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PV6 | |
LENG8 | |
Recombinant protein corresponding to Human LENG8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only