missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leishman Stain
26.98 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.
Solubility
Soluble in water (4 mg/ml), methanol (1 mg/ml), ethanol (4 mg/ml), and methyl cellosolve (20 mg/ml).
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Leishman Stain is used to stain blood smears to identify parasites, leukocytes and trypanosomas. It is used in hematology and tropical medicine to differentiate and identify leukocytes. It acts like fluorescent stains for detection of malaria parasite. It is also used for bone marrow staining.
Solubility
Soluble in water (4 mg/ml), methanol (1 mg/ml), ethanol (4 mg/ml), and methyl cellosolve (20 mg/ml).
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Solubility Information | Soluble in water (4mg/ml),methanol (1mg/ml),ethanol (4mg/ml),and methyl cellosolve (20mg/ml). |
Odor | Odorless |
Quantity | 25 g |
Chemical Name or Material | Leishman Stain |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'