missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Lefty Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Lefty |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Lefty Polyclonal specifically detects Lefty in Human, Mouse samples. It is validated for Western Blot.Specifications
Lefty | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse | |
LEFTY2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q64280 | |
Mouse myeloma cell line NS0-derived recombinant mouse Lefty-1 Leu22-Arg135, His6, Leu136-Pro368 Accession # Q64280 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human and mouse Lefty in direct ELISAs and Western blots. |
For Research Use Only