Learn More
LECT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560541
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NKNAINNGVR ISGRGFCVKM FYIKPIKYKG PIKKGEKLGT LLPLQKVYPG IQSHVHIENC DSSDPT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
This gene encodes a secreted, 16 kDa protein that acts as a chemotactic factor to neutrophils and stimulates the growth of chondrocytes and osteoblasts. This protein has high sequence similarity to the chondromodulin repeat regions of the chicken myb-induced myeloid 1 protein. A polymorphism in this gene may be associated with rheumatoid arthritis.Specifications
LECT2 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14960 | |
LECT2 | |
Recombit protein corresponding to Human LECT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
LECT2 | |
Leukocyte cell-derived chemotaxin 2; hLECT2; LECT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only