missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LECT2 Mouse anti-Human, Clone: 102717, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | LECT2 |
---|---|
Clone | 102717 |
Immunogen | E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 |
Gene ID (Entrez) | 3950 |
Gene Accession No. | O14960 |
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.Specifications
LECT2 | |
E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 | |
O14960 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG1 | |
Protein A or G purified from ascites | |
RUO |
102717 | |
3950 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects recombinant human LECT2 in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only