missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LECT2 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | LECT2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
LECT2 Polyclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.Specifications
LECT2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1 | |
LECT2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O14960 | |
3950 | |
E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human LECT2 in direct ELISAs and Western blots. |
For Research Use Only