missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead wire, 0.75mm (0.03in) dia, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 m
2 m
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Gray |
Quantity | 2 m |
Percent Purity | 99.995% |
Odor | Odorless |
Weight | ≈5g/m |
Form | Wire |
Assay | (metals basis) |
Chemical Name or Material | Lead wire, 0.75mm (0.03 in.) dia. |
Safety and Handling
Product Identifier
- Lead wire
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'