missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Brand:  Alfa Aesar™ 041025.TB

Code : NEW

Additional Details : Weight : 0.11300kg

Quantity 113g

 View more versions of this product

Product Code. 11309437

  • £26.92 / 113g


Estimated Shipment date
from Supplier 19-01-2018
Add to basket

Description

Description

  • For use with copper, copper alloys, lead, steel, and nickel alloys
  • Recommended for machine and torch soldering
Specifications

Chemical Identifiers

Lead Tin, Solder Alloy
3.18mm (0.125 in.) dia.
MFCD00798549

Specifications

Pb:Sn; 70:30 wt%
113g
MFCD00798549
Lead Tin, Solder Alloy
3.18mm (0.125 in.) dia.
Videos
SDS