missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 041025.29

Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Code : NEW

Additional Details : Weight : 0.22500kg

Product Code. 11399427

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.