missing translation for 'onlineSavingsMsg'
Learn More

Lead, Technical, SLR, Metal Shot, about 2mm, Fisher Chemical™

Catalog Number 10121530
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]
This item is not returnable. View return policy
7439-92-1
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
5352425
CHEBI:27889
lead
[Pb]
Color Gray
Melting Point 327.4°C
Boiling Point 1740°C
Quantity 1 kg
Formula Weight 207.19g/mol
Packaging Plastic powder jar
Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'