missing translation for 'onlineSavingsMsg'
Learn More

Lead subacetate, ACROS Organics™

Brand:  Acros Organics 217540010

Code : 29

Additional Details : CAS Number : 1335-32-6 Weight : 1.05000kg Transport : UN number : 1616 Chem class : 6.1 Pack group : III


Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10411071

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Lead subacetate
1335-32-6
≥72% (Pb)
(CH3CO2)2Pb·2Pb(OH)2
MFCD00013041
c.p.
Extra Pure
C4H10O8Pb3
807.69
Basic lead acetate

Specifications

Lead subacetate
1335-32-6
Granular Powder
C4H10O8Pb3
807.69
15, 5474
300ppm max.
Authentic
200ppm max.
Plastic bottle
White
c.p.
Extra Pure
≥72% (Pb)
(CH3CO2)2Pb·2Pb(OH)2
MFCD00013041
Basic lead acetate
200ppm max.
0.05% max. (in dilute acetic acid)
200ppm max.
Solubility in water: 62.5g/L. Other solubilities: 250g/L in hot water, soluble in dilute acetic acid
1kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lead subacetate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS