missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

Catalog Number p-218846
Click to view available options
Quantity:
30 mL
125 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7439-92-1, 7697-37-2, 7732-18-5
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
CHEBI:27889
lead
[Pb]
This item is not returnable. View return policy
7439-92-1, 7697-37-2, 7732-18-5
Pb
207.20
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
CHEBI:27889
lead
[Pb]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Grade Trace ICP/ICP-MS
Packaging Triple-leached LDPE bottle
Solvent or Matrix 2% HNO3
Quantity 30 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Lead Standard
compliance-icons compliance-icons
Product Identifier
  • 1000µg/ml lead
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, lead

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'