missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, 10,000μg/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep CL-CRDL-1

Code : 04

Additional Details : Weight : 0.09000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 500mL
Packaging Triple-leached LDPE bottle

 View more versions of this product

Product Code. 10661236

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet





Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001


GHS/OSHA Complaint


Chemical Identifiers

Lead Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation


Lead Standard
Matrix&tab;Concentration&tab;Cat. No.
2-5% HNO3&tab;10, 000μg/mL&tab;PLPB2-3Y, -3X
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
5% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Plasma Grade
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.

Product intended for laboratory use only