missing translation for 'onlineSavingsMsg'
Learn More

Lead rod, 6.35mm (0.25in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 15406545
Click to view available options
Quantity:
1 m
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
CAS 7439-92-1
Molecular Formula Pb
MDL Number MFCD00134050
Quantity 1 m
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES [Pb]
IUPAC Name lead
Molecular Weight (g/mol) 207.20
PubChem CID 5352425
ChEBI CHEBI:27889
Percent Purity 99.95%
Odor Odorless
Weight ≈3.6g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Lead rod, 6.35mm (0.25 in.) dia.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead rod
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'