missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Oxide, BAKER ANALYZED™ Reagent, J.T. Baker™
167.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Quantity | 500 g |
Formula Weight | 223.19 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 9.53kg |
Chemical Name or Material | Lead Oxide |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P281-Use personal protective equipment as required.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'