missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead Nitrate Solution, EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Specifications
Quantity | 1000 mL |
Safety and Handling
Product Identifier
- Lead nitrate 0.05M Volumetric Solution European Pharmacopoeia Ref: 3009700
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH201A-Warning
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'