Learn More
Thermo Scientific Alfa Aesar Lead naphthenate, 61% min in mineral spirits, Pb 20%
Brand: Thermo Scientific Alfa Aesar 038593.30
Lead naphthenate, 61% min in mineral spirits, Pb 20%
Code : NEW
Additional Details : CAS Number : 61790-14-5 Weight : 0.25000kg Transport : UN number : 1993 Chem class : 3 Pack group : III