missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead naphthenate, 61% min in mineral spirits, Pb 20%

Brand:  Alfa Aesar™ 038593.A1

Code : NEW

Additional Details : CAS Number : 61790-14-5 Weight : 1.00000kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 1kg

 View more versions of this product

Product Code. 11338907

  • £130.20 / 1kg


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Specifications

Chemical Identifiers

Lead naphthenate
61790-14-5
MFCD00466946
UN1993

Specifications

Lead naphthenate
61790-14-5
1.13g/mL
40° (104°F)
MFCD00466946
1kg
61% min. in min.eral spirits, Pb 20%
Brown
Liquid
UN1993
Videos
SDS