Learn More
Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals
Description
It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 290°C (decomposition) |
Color | Brown to Black |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 97.0% min. |
Solubility Information | Insoluble in water and alcohol. Soluble in acetic acid. |
IUPAC Name | dioxolead |
Formula Weight | 239.19 |
Grade | ACS |
Odor | Odorless |
Show More |
RUO – Research Use Only