missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead in Isooctane standards, complete set for method ASTM D5059 Part C

Complete set for method ASTM D5059 Part C

Brand:  Alfa Aesar™ 045337.KS

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1262 Chem class : 1 Pack group : II


Quantity 1 each

Product Code. 15406915

  • £477.60 / Each


Estimated Shipment date
from Supplier 18-05-2018
Add to basket


Specifications

Specifications

Lead in Isooctane standards
MFCD00134050
Matrix: Isooctane
1 each
Liquid
UN1262
Colorless
Videos
SDS