missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead in Isooctane standard solution, Specpure™, 111μg/g(0.300g/gal)

Brand:  Alfa Aesar™ 045336.22

Code : NEW

Additional Details : Weight : 0.10000kg Transport : UN number : 1262 Chem class : 1 Pack group : II


Quantity 100g

Product Code. 15496905

  • £88.90 / 100g


Estimated Shipment date
from Supplier 22-03-2018
Add to basket


Specifications

Specifications

Lead in Isooctane standard solution
Specpure™
MFCD01094200
Nearly odorless
100g
111μg/g(0.300g/gal)
Liquid
UN1262
Colorless
Matrix: Isooctane
Videos
SDS