Learn More
Alfa Aesar™ Lead(II) zirconium(IV) 2-ethylhexanoate tetraisopropoxide, 99% (metals basis), 10% w/v in hexane
Soluble precursor of lead zirconium mixed oxide
Brand: Alfa Aesar™ 036575.AC
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1208 Chem class : 1 Pack group : II
Quantity | 25mL |
---|
Chemical Identifiers
Lead(II) zirconium(IV) 2-ethylhexanoate tetraisopropoxide | |
MFCD00171337 | |
Lead zirconium 2-ethylhexano-isopropoxide |
821.18 | |
UN1208 |
Specifications
Lead(II) zirconium(IV) 2-ethylhexanoate tetraisopropoxide | |
10% w/v in hexane | |
Moisture Sensitive | |
99% | |
821.18 | |
UN1208 |
25mL | |
Brown | |
Liquid | |
(metals basis) | |
MFCD00171337 | |
Lead zirconium 2-ethylhexano-isopropoxide |