missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead(II) thiocyanate, 99.99% (metals basis)

Brand:  Alfa Aesar™ 044084.14

Code : NEW

Additional Details : CAS Number : 592-87-0 Weight : 0.02500kg Transport : UN number : 2291 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11385178

  • £50.99 / 25g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Lead (II) thiocyanate
Pb(SCN)2
MFCD00011155
3687984
592-87-0
323.35
UN2291
14,5426

Specifications

Lead (II) thiocyanate
Powder
(metals basis)
323.35
UN2291
14,5426
Odorless
25g
592-87-0
99.99%
Pb(SCN)2
MFCD00011155
3687984
3.82g/mL
190°C
Videos
SDS