Learn More
Lead(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals
Description
Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 3 x 1 kg |
Solubility Information | Miscible with water. |
IUPAC Name | λ²-lead(2+) bis(tetrafluoroboranuide) |
Formula Weight | 380.81 |
Odor | Odorless |
Density | 1.62 |
Chemical Name or Material | Lead(II) tetrafluoroborate |
RUO – Research Use Only