missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 15432287
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
1314-87-0
PbS
239.30
MFCD00016280
FILXVLQXZYXBSJ-UHFFFAOYSA-N
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
14819
[S--].[Pb++]
This item is not returnable. View return policy
1314-87-0
PbS
239.30
MFCD00016280
FILXVLQXZYXBSJ-UHFFFAOYSA-N
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
14819
[S--].[Pb++]

Lead(II) sulfide is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1114°C
Color Black
Physical Form Lump
Quantity 10 g
Assay Percent Range 99.995% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name λ²-lead(2+) sulfanediide
Formula Weight 239.25
Odor Odorless
Refractive Index 3.921
Appearance Black lumps
Density 7.5
Chemical Name or Material Lead(II) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'