Learn More
Alfa Aesar™ Lead(II) selenate, 99.9%
Brand: Alfa Aesar™ 039345.06
Code : NEW
Additional Details : CAS Number : 7446-15-3 Weight : 0.00500kg Transport : UN number : 2630 Chem class : 1 Pack group : I
Quantity | 5g |
---|
Chemical Identifiers
Lead(II) selenate | |
350.16 | |
UN2630 |
7446-15-3 | |
MFCD00054013 |
Specifications
Lead(II) selenate | |
Powder | |
350.16 | |
UN2630 | |
6.37g/mL |
7446-15-3 | |
99.9% | |
MFCD00054013 | |
Insoluble in water; Soluble in concentrated acids | |
5g |