missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead(II) selenate, 99.9%

Brand:  Alfa Aesar™ 039345.14

Code : NEW

Additional Details : CAS Number : 7446-15-3 Weight : 0.02500kg Transport : UN number : 2630 Chem class : 6.1 Pack group : I

Quantity 25g

 View more versions of this product

Product Code. 11376489

  • £220.48 / 25g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

Lead(II) selenate
350.16
UN2630
7446-15-3
MFCD00054013

Specifications

Lead(II) selenate
Powder
350.16
UN2630
6.37g/mL
7446-15-3
99.9%
MFCD00054013
Insoluble in water; Soluble in concentrated acids
25g
Videos
SDS