missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-7048842
Click to view available options
Quantity:
100 g
500 g
13453-62-8
Cl2H6O11Pb
460.10
MFCD00149817
KKGGEAQRICYXNM-UHFFFAOYSA-L
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
25021834
lead(2+);diperchlorate;trihydrate
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
13453-62-8
Cl2H6O11Pb
460.10
MFCD00149817
KKGGEAQRICYXNM-UHFFFAOYSA-L
lead ii perchlorate trihydrate, acmc-20aknd, 2clo4.pb.3h2o, lead perchlorate 3h2o, lead ii perchlorate, trihydrate, lead perchlorate, trihydrate, reagent acs, lead ii perchlorate trihydrate, acs reagent
25021834
lead(2+);diperchlorate;trihydrate
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Titrant in complexometric titrations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100°C (decomposition)
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Pb(ClO4)2·3H2O
UN Number UN1470
Solubility Information Soluble in water and alcohol.
Formula Weight 460.15 (406.10 Anhydrous)
Percent Purity ≤97%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 2.60 g/mL
Chemical Name or Material Lead(II) perchlorate trihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead (II) perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P283-Wear fire/flame resistant/retardant clothing.
  • P306+P360-IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'