missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 1470°C |
Melting Point | 890°C |
Color | Yellow |
Physical Form | Solid |
Quantity | 50 g |
Linear Formula | PbO |
UN Number | UN2291 |
IUPAC Name | oxolead |
Formula Weight | 223.2g/mol |
Percent Purity | ≥99.0% (KT) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'