missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead(II) oxalate, Puratronic™, 99.999% (metals basis)

Brand:  Alfa Aesar™ 012999.14

UNSPSC : 12141723

Code : NEW

Additional Details : CAS Number : 814-93-7 Weight : 0.01000kg Transport : UN number : 2291 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11342098

  • £95.72 / 25g


Estimated Shipment date
from Supplier 13-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Lead(II) oxalate
Puratronic™
295.23
UN2291
814-93-7
PbC2O4
MFCD00040677

Specifications

295.23
MFCD00040677
(metals basis)
PbC2O4
Odorless
Lead(II) oxalate
99.999%
Puratronic™
UN2291
25g
300°C (dec.)
Powder
814-93-7
Documentation

Technical Documents