missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) carbonate, basic, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Lead(II) carbonate is employed in the production of anticorrosion paint and outdoor paint. It is also used as a precursor for cosmetics, colored glaze and dope. It finds application in the tin-lead plating. It is also useful as a stabilizing agent for vinyl chloride plastic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 400°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99% (Metal Basis) |
UN Number | 1479 |
Synonym | White L ead, C.I. 77597 |
Solubility Information | Slightly soluble in carbon dioxide solution,acetic and dilute nitric acid. Insoluble in water and ethanol. |
Formula Weight | 775.63 |
Appearance | White powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'