missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) acetate, basic, ACS, Thermo Scientific Chemicals
52.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
Lead(II) acetate is used in the production of sweeteners, cosmetics, astringent, used in the cleaning and maintenance of stainless steel firearm suppressors (silencers) and compensators, and also used in making of slow matches.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | Pb(OOCCH3)2·2Pb(OH)2 |
UN Number | UN1616 |
Merck Index | 14,5419 |
Solubility Information | Soluble in water. |
Formula Weight | 807.71 |
Grade | ACS |
Sensitivity | Air Sensitive |
Chemical Name or Material | Lead(II) acetate, Basic |
Safety and Handling
Product Identifier
- Lead(II) acetate, basic
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H351-Suspected of causing cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'