missing translation for 'onlineSavingsMsg'
Learn More

Lead Granules, +30 mesh, 99.99% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 045081.A4

Code : NEW

Additional Details : CAS Number : 7439-92-1 Weight : 2.50000kg

Quantity 2.5kg

 View more versions of this product

Product Code. 15456725

  • £119.60 / 2.50kg


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Lead
Granular
MFCD00134050
7439-92-1
99.99% (Metals basis)

Specifications

Lead
7439-92-1
Granular
MFCD00134050
Gray
2.5kg
Gray granules
99.99% (Metals basis)
Odorless
Videos
SDS