missing translation for 'onlineSavingsMsg'
Learn More

Lead citrate, trihydrate, 96%, Thermo Scientific™

Catalog Number p-153287
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6107-83-1
C12H16O17Pb3
1053.843
MFCD00150456
GMPMGSCJCDAUMP-UHFFFAOYSA-H
lead ii citrate trihydrate, acmc-20akng, citric acid lead ii salt trihydrate, tris ? 2-lead 2+ dicitrate trihydrate, 2-hydroxypropane-1,2,3-tricarboxylate; lead 2+ ; trihydrate, 1,2,3-propanetricarboxylicacid, 2-hydroxy-, lead 2+ salt, hydrate 2:3:3
2735068
2-hydroxypropane-1,2,3-tricarboxylate;lead(2+);trihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Pb+2].[Pb+2].[Pb+2]
This item is not returnable. View return policy
6107-83-1
C12H16O17Pb3
1053.843
MFCD00150456
GMPMGSCJCDAUMP-UHFFFAOYSA-H
lead ii citrate trihydrate, acmc-20akng, citric acid lead ii salt trihydrate, tris ? 2-lead 2+ dicitrate trihydrate, 2-hydroxypropane-1,2,3-tricarboxylate; lead 2+ ; trihydrate, 1,2,3-propanetricarboxylicacid, 2-hydroxy-, lead 2+ salt, hydrate 2:3:3
2735068
2-hydroxypropane-1,2,3-tricarboxylate;lead(2+);trihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Pb+2].[Pb+2].[Pb+2]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (HOC(COO-)(CH2COO-)2)2Pb3·3H2O
Quantity 100 g
Formula Weight 1053.85
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Lead citrate, trihydrate, 96%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead citrate, trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'