missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead Antimony shot, 2mm (0.08in)

Brand:  Alfa Aesar™ 010224.A3

Code : NEW

Additional Details : CAS Number : 7439-92-1 Weight : 2.00000kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 2kg

 View more versions of this product

Product Code. 11301288

  • £64.40 / 2kg


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Specifications

Chemical Identifiers

Lead Antimony shot
2mm (0.08 in.)
MFCD00134050
UN3077

Specifications

Lead Antimony shot
2mm (0.08 in.)
MFCD00134050
Odorless
2kg
Shot
UN3077
Videos
SDS