missing translation for 'onlineSavingsMsg'
Learn More

Brand™ LDPE Wash Bottles with Venting Valve

High-quality wash bottles with integrated wash stream and sealing cone closure.

Brand:  Brand™ 144028

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 10761383

  • £38.51 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • For distilled water and organic solvents only
  • Large opening for convenient filling
  • Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
  • Manufactured from flexible PE-LD, limited to temperatures below 80°C

Specifications

Specifications

500mL
Ethanol
Wash bottle
Orange
Stopper
500mL
LDPE
Yes
Wide
Videos