Learn More
Lazabemide hydrochloride, Tocris Bioscience™
Selective MAO-B inhibitor
Brand:  Tocris (Bio-Techne) 246050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13461254
  • £414.72 / 50mg
Estimated Shipment
03-09-2018
Add to basket
Description
Description
Selective, reversible monoamine oxidase B (MAO-B) inhibitor (IC50 values are 0.03 and > 100 μM for MAO-B and MAO-A respectively). Inhibits monoamine uptake at high concentrations (IC50 values are 86, 123 and > 500 μM for noradrenalin, serotonin and dopamine uptake respectively).
Specifications
Chemical Identifiers
103878-83-7
236.096
N-(2-Aminoethyl)-5-chloro-2-pyridinecarboxamide hydrochloride
C8H11Cl2N3O
JMFKTFLARGGXCC-UHFFFAOYSA-N
163727
Specifications
Lazabemide hydrochloride
>98%
N-(2-Aminoethyl)-5-chloro-2-pyridinecarboxamide hydrochloride
C1=CC(=NC=C1Cl)C(=O)NCCN.Cl
236.096
236.1
103878-83-7
C8H11Cl2N3O
JMFKTFLARGGXCC-UHFFFAOYSA-N
N-(2-aminoethyl)-5-chloropyridine-2-carboxamide;hydrochloride
163727