missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Layilin Ectodomain Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Layilin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Layilin Polyclonal specifically detects Layilin in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Layilin | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
LAYN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q8C351 | |
143903 | |
Mouse myeloma cell line NS0-derived recombinant mouse Layilin Ectodomain Lys18-Glu229 (Ala29-Gly36 del) Accession # Q8C351 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Layilin Ectodomain in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant human Layilin is observed. |
For Research Use Only