Learn More
LAT, clone: LAT.10-17 (10-17), eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-9967-80
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15278287
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
This LAT.10-17 (10-17) monoclonal antibody reacts with human Linker for Activation of T cells (LAT), which is essential for T cell receptor (TCR)-mediated signal transduction. Characterized as an integral transmembrane protein, this ∽37kDa molecule does not possess a true extracellular domain. Upon TCR/CD3 engagement, LAT localizes to lipid rafts and is tyrosine phosphorylated by ZAP-70, leading to the recruitment and activation of other downstream signaling molecules such as Grb-2, PLCγ, and PI3K. This adapter molecule has been shown to be required for T cell activation, as well as thymocyte and T regulatory cell development. In humans, LAT is expressed on peripheral blood lymphocytes, lymph nodes, and tonsil. Studies in mice have shown that LAT is expressed in thymocytes, T cells, mast cells, natural killer cells, megakaryocytes, platelets, and early B cells. This antibody does not crossreact with the mouse antigen.
Specifications
Specifications
LAT
LAT.10-17 (10-17)
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
ImmunoprecipitationþWestern Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Linker for activation of T-cells
IgG1, kappa