Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250μL and larger.
Brand:  Hamilton™ 7806-07
Code : A0
Additional Details : Weight : 0.50000kg
Product Code. 13498736
  • £146.00 / Pack of 6
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs