Learn More
CleanSpace Technology™ Large Case Particulate Pre-Filters
Suitable for protection against airborne particulates (dust, mists and fumes). Designed to remove course particles and enhance the life of the Particulate P3 Filter on PAF-0037 and Combined Filters. The Pre-Filter should be changed after each use.
Brand:  CleanSpace Technology™ PAF-0057
Additional Details : Weight : 0.05200kg
Product Code. 15980062
  • £39.65 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Used with the revolutionary CleanSpace - a light weight PAPR with no hoses/belts
  • Materials: Spun polymer fibers
  • Easy and quickly fitted and removed from the power unit
  • Sold in packs of 20
  • Compatible with all CleanSpace Respirators
Specifications
Specifications
Prefilter